Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLX Antibody (PCRP-MLX-1G8), PerCP, Novus Biologicals™
SDP

Catalog No. NB382487 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382487 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382487 Supplier Novus Biologicals Supplier No. NBP308241PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MLX Monoclonal antibody specifically detects MLX in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen MLX
Applications Western Blot, Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-MLX-1G8
Conjugate PerCP
Formulation PBS
Gene Alias BHLHD13, bHLHd13Max-like bHLHZip protein, BigMax protein, Class D basic helix-loop-helix protein 13, MAD7, MAX-like protein X, MXD7, TCFL4Protein BigMax, transcription factor-like 4, Transcription factor-like protein 4
Host Species Mouse
Immunogen Recombinant full-length human MLX protein (Uniprot: Q9UH92)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6945
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.