Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MLXIPL Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501728607 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8607 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8607 Supplier Proteintech Group Inc Supplier No. 132561AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ChREBP is a transcription factor involved in activating genes that encode enzymes of fatty acid biosynthesis in liver and adipose. This protein may have relevance to lipid metabolism, obesity, and type II diabetes.

Specifications

Antigen MLXIPL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MLXIPL
Gene Accession No. Q9NP71
Gene Alias bHLHd14, CHREBP, MIO, MLX interacting protein like, MLX interactor, MLXIPL, MONDOB, WBSCR14, WS bHLH
Gene Symbols MLXIPL
Host Species Rabbit
Immunogen MLXIPL Fusion Protein Ag3481
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51085
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.