Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMAB Antibody (OTI2G5), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP272730A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272730A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272730A94 Supplier Novus Biologicals Supplier No. NBP272730AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMAB Monoclonal specifically detects MMAB in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen MMAB
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI2G5
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias aquocob(I)alamin vitamin B12s adenosyltransferase, ATP:cob(I)alamin adenosyltransferase, ATP:corrinoid adenosyltransferase, ATR, cblB, c-diamide adenosyltransferase, mitochondrial, cob, Cob(I)alamin adenosyltransferase, cob(I)yrinic acid a, EC 2.5.1.17, methylmalonic aciduria (cobalamin deficiency) cblB type, methylmalonic aciduria (cobalamin deficiency) type B, Methylmalonic aciduria type B protein, MGC20496
Gene Symbols MMAB
Host Species Mouse
Immunogen Full length recombinant protein of human MMAB (NP_443077) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 326625.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.