Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMADHC Antibody (OTI1G4), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP272739IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272739IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272739IR Supplier Novus Biologicals Supplier No. NBP272739IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMADHC Monoclonal specifically detects MMADHC in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen MMADHC
Applications Western Blot
Classification Monoclonal
Clone OTI1G4
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cblD, chromosome 2 open reading frame 25, methylmalonic aciduria (cobalamin deficiency) cblD type, with homocystinuria, mitochondrial, protein C2orf25, mitochondrial
Gene Symbols MMADHC
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 39-296 of human MMADHC (NP_056517) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 27249.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.