Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMD2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15638820 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15638820 20 μL
NBP156388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15638820 Supplier Novus Biologicals Supplier No. NBP15638820UL

Rabbit Polyclonal Antibody

MMD2 Polyclonal specifically detects MMD2 in Human samples. It is validated for Western Blot.

Specifications

Antigen MMD2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q8IY49
Gene Alias FLJ37205, monocyte to macrophage differentiation factor 2, monocyte to macrophage differentiation-associated 2, monocyte-to-macrophage differentiation factor 2, PAQR10Progestin and adipoQ receptor family member 10, Progestin and adipoQ receptor family member X
Gene Symbols MMD2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to MMD2(monocyte to macrophage differentiation-associated 2) The peptide sequence was selected from the N terminal of MMD2. Peptide sequence FAPRLLDFQKTKYARFMNHRVPAHKRYQPTEYEHAANCATHAFWIIPSIL.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221938
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.