Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-10 Antibody (MM0024-9G10) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB017987 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017987 0.025 mg
NB11060990 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017987 Supplier Novus Biologicals Supplier No. NB110609900.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMP-10 Monoclonal specifically detects MMP-10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MMP-10
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0019-9G10
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:300
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Alias EC 3.4.24, EC 3.4.24.22, matrix metallopeptidase 10 (stromelysin 2), matrix metalloprotease 10, matrix metalloproteinase 10 (stromelysin 2), Matrix metalloproteinase-10, MMP-10, SL-2, STMY2stromelysin-2, stromelysin 2, transin 2, transin-2
Gene Symbols MMP10
Host Species Mouse
Immunogen This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified human recombinant MMP-10).
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4319
Test Specificity MMP10 (MM0019-9G10)
Reconstitution Reconstitute with 0.5 ml sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.