Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-10 Antibody (MM0024-9G10), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB362419 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362419 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362419 Supplier Novus Biologicals Supplier No. NB11060990MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMP-10 Monoclonal antibody specifically detects MMP-10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen MMP-10
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0024-9G10
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.24, EC 3.4.24.22, matrix metallopeptidase 10 (stromelysin 2), matrix metalloprotease 10, matrix metalloproteinase 10 (stromelysin 2), Matrix metalloproteinase-10, MMP-10, SL-2, STMY2stromelysin-2, stromelysin 2, transin 2, transin-2
Host Species Mouse
Immunogen Human recombinant MMP-10
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4319
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.