Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-11 Antibody (135421), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB371774 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371774 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371774 Supplier Novus Biologicals Supplier No. FAB3657I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMP-11 Monoclonal specifically detects MMP-11 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining).

Specifications

Antigen MMP-11
Applications CyTOF, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 135421
Conjugate Janelia Fluor 549
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.24, EC 3.4.24.-, EC 3.4.24.24, matrix metallopeptidase 11 (stromelysin 3), matrix metalloproteinase 11 (stromelysin 3), Matrix metalloproteinase-11, MMP-11, SL-3, ST3, STMY3stromelysin 3, stromelysin III, stromelysin-3
Host Species Mouse
Immunogen E. coli-derived recombinant human MMP-11, Met1-Leu488, Accession # P24347
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4320
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.