Learn More
Description
Specifications
Specifications
| Antigen | MMP-12 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 701E4.03 |
| Concentration | 0.5mg/mL |
| Conjugate | HRP |
| Dilution | ELISA |
| Formulation | PBS and 50% Glycerol with No Preservative |
| Gene Alias | EC 3.4.24, hME, HMEEC 3.4.24.65, Macrophage elastase, macrophage metalloelastase, matrix metallopeptidase 12 (macrophage elastase), matrix metalloproteinase 12 (macrophage elastase), Matrix metalloproteinase-12, ME, MGC138506, MME, MMP-12 |
| Gene Symbols | MMP12 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
