Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-12 Antibody (706F9.01), HRP, Novus Biologicals™
SDP

Catalog No. DDX0281HRPO Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0281HRPO 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0281HRPO Supplier Novus Biologicals Supplier No. DDX0281HRPO
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

MMP-12 Monoclonal specifically detects MMP-12 in Human samples. It is validated for ELISA.

Specifications

Antigen MMP-12
Applications ELISA
Classification Monoclonal
Clone 706F9.01
Conjugate HRP
Dilution ELISA 1:100-1:2000
Gene Accession No. NP_002417
Gene Alias EC 3.4.24, hME, HMEEC 3.4.24.65, Macrophage elastase, macrophage metalloelastase, matrix metallopeptidase 12 (macrophage elastase), matrix metalloproteinase 12 (macrophage elastase), Matrix metalloproteinase-12, ME, MGC138506, MME, MMP-12
Gene Symbols MMP12
Host Species Mouse
Immunogen MMP-12/HME was used as immunogen.
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4321
Target Species Human
Content And Storage Store at -20C in the dark. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.