Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-13 Antibody (OTI2D8), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N272740J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272740J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272740J549 Supplier Novus Biologicals Supplier No. NBP272740JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMP-13 Monoclonal specifically detects MMP-13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.

Specifications

Antigen MMP-13
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Monoclonal
Clone OTI2D8
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP-13
Gene Symbols MMP13
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 104-471 of human MMP-13 (NP_002418) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4322.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.