Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-13 Antibody (VIIIA2), Novus Biologicals™
SDP

Catalog No. NB1105919 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1105919 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1105919 Supplier Novus Biologicals Supplier No. NB1105919
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMP-13 Monoclonal specifically detects MMP-13 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen MMP-13
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone VIIIA2
Concentration 0.4 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 - 4 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:25 - 1:50, Immunohistochemistry-Paraffin 1:12.5 - 1:25
Gene Accession No. P45452
Gene Alias CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP-13
Gene Symbols MMP13
Host Species Mouse
Immunogen Human recombinant collagenase-3 protein
Purification Method Protein G purified
Quantity 500 μL
Regulatory Status RUO
Research Discipline Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4322
Test Specificity Recognizes pro and acive forms of MMP 13.
Target Species Human, Mouse, Rat, Rabbit
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.