Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-14/MT1-MMP Antibody (128527), FITC, Novus Biologicals™
SDP

Catalog No. NB295510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295510 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295510 Supplier Novus Biologicals Supplier No. FAB9181F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMP-14/MT1-MMP Monoclonal specifically detects MMP-14/MT1-MMP in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen MMP-14/MT1-MMP
Applications CyTOF, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 128527
Conjugate FITC
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF-ready
Formulation PBS
Gene Alias EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP-X1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MMP-14/MT1-MMP, Tyr112-Ala541 (predicted), Accession # P50281
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4323
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.