Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-14/MT1-MMP Antibody (3G4.2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB019446 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019446 0.025 mg
NBP229735 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019446 Supplier Novus Biologicals Supplier No. NBP2297350.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMP-14/MT1-MMP Monoclonal specifically detects MMP-14/MT1-MMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MMP-14/MT1-MMP
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 3G4.2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500
Formulation PBS (pH 7.2) with 0.1% Sodium Azide
Gene Accession No. P50281
Gene Alias EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP-X1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1
Gene Symbols MMP14
Host Species Mouse
Immunogen Recombinant protein containing the catalytic domain of human MT1-MMP.
Molecular Weight of Antigen 60 kDa
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4323
Test Specificity Clone 3G4.2 recognizes the catalytic domain of MT1-MMP, identifying the active form of the protein. MW is approximately 60 kDa.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.