Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-14/MT1-MMP Antibody (MM0027-9E10), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB160993646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160993646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB160993646 Supplier Novus Biologicals Supplier No. NB11060993JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMP-14/MT1-MMP Monoclonal specifically detects MMP-14/MT1-MMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MMP-14/MT1-MMP
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0027-9E10
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.4.24, EC 3.4.24.80, matrix metallopeptidase 14 (membrane-inserted), matrix metalloproteinase 14 (membrane-inserted), matrix metalloproteinase-14, membrane type 1 metalloprotease, Membrane-type matrix metalloproteinase 1, Membrane-type-1 matrix metalloproteinase, MMP-14, MMP-X1, MT1MMP, MT1-MMPMTMMP1, MT-MMP 1
Gene Symbols MMP14
Host Species Mouse
Immunogen Human recombinant MMP-14/MT1-MMP
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4323
Test Specificity This antibody recognizes human MMP-14 in western blot without cross reactivity with human MMP-1-13, and MMP14-15.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.