Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-15/MT2-MMP Antibody (130522), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB295530 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295530 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295530 Supplier Novus Biologicals Supplier No. FAB9161T

Mouse Monoclonal Antibody

MMP-15/MT2-MMP Monoclonal antibody specifically detects MMP-15/MT2-MMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF

Specifications

Antigen MMP-15/MT2-MMP
Applications Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 130522
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 15 (membrane-inserted), matrix metalloproteinase 15 (membrane-inserted), Membrane-type matrix metalloproteinase 2, Membrane-type-2 matrix metalloproteinase, MMP-15, MT2-MMPMT2MMP, MTMMP2MT-MMP 2, SMCP-2matrix metalloproteinase-15
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MMP-15, aa 132-625
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4324
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.