Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-15/MT2-MMP Antibody (MM0028-5D11), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBT60994647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBT60994647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBT60994647 Supplier Novus Biologicals Supplier No. NB11060994AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMP-15/MT2-MMP Monoclonal specifically detects MMP-15/MT2-MMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MMP-15/MT2-MMP
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0028-5D11
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 15 (membrane-inserted), matrix metalloproteinase 15 (membrane-inserted), Membrane-type matrix metalloproteinase 2, Membrane-type-2 matrix metalloproteinase, MMP-15, MT2-MMPMT2MMP, MTMMP2MT-MMP 2, SMCP-2matrix metalloproteinase-15
Gene Symbols MMP15
Host Species Mouse
Immunogen Human recombinant MMP-15/MT2-MMP
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4324
Test Specificity This antibody recognizes human MMP-15 in western blot without cross reactivity with human MMP-14/16.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.