Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-16/MT3-MMP Antibody (782005), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB349620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349620 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349620 Supplier Novus Biologicals Supplier No. FAB1785MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMP-16/MT3-MMP Monoclonal specifically detects MMP-16/MT3-MMP in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen MMP-16/MT3-MMP
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 782005
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias chromosome 8 open reading frame 57, DKFZp761D112, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 16 (membrane-inserted), matrix metalloproteinase 16 (membrane-inserted), matrix metalloproteinase-16, Membrane-type matrix metalloproteinase 3, Membrane-type-3 matrix metalloproteinase, MMP-16, MMPX2, MMP-X2, MT3MMP, MT3-MMPC8orf57, MT-MMP 3, MT-MMP2, MTMMP3, MT-MMP3, Putative transmembrane protein C8orf57
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human MMP-16/MT3-MMP, Ala32-Pro535, Accession # P51512
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4325
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.