Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-19 Antibody, Novus Biologicals™
SDP

Catalog No. NBP217311 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP217311 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP217311 Supplier Novus Biologicals Supplier No. NBP217311
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MMP-19 Polyclonal specifically detects MMP-19 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.

Specifications

Antigen MMP-19
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, ELISA, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Knockdown Validated
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Alias EC 3.4.24.-, matrix metallopeptidase 19, matrix metalloproteinase 18, matrix metalloproteinase 19, Matrix metalloproteinase RASI, Matrix metalloproteinase-18, matrix metalloproteinase-19, MMP-18, MMP18MMP-19, RASI, RASI-1
Gene Symbols MMP19
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MMP19. The exact sequence is proprietary.
Molecular Weight of Antigen 57 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4327
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.