Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-19 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15709420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15709420 20 μL
NBP157094 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15709420 Supplier Novus Biologicals Supplier No. NBP15709420UL

Rabbit Polyclonal Antibody

MMP-19 Polyclonal specifically detects MMP-19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MMP-19
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q99542
Gene Alias EC 3.4.24.-, matrix metallopeptidase 19, matrix metalloproteinase 18, matrix metalloproteinase 19, Matrix metalloproteinase RASI, Matrix metalloproteinase-18, matrix metalloproteinase-19, MMP-18, MMP18MMP-19, RASI, RASI-1
Gene Symbols MMP19
Host Species Rabbit
Immunogen Synthetic peptides corresponding to MMP19 (matrix metallopeptidase 19) The peptide sequence was selected from the C terminal of MMP19. Peptide sequence IHFFKGDKVWRYINFKMSPGFPKKLNRVEPNLDAALYWPLNQKVFLFKGS.
Purification Method Protein A purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4327
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.