Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-2 Antibody (1A10), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB174459 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174459 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174459 Supplier Novus Biologicals Supplier No. FAB903E
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMP-2 Monoclonal specifically detects MMP-2 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen MMP-2
Applications CyTOF, Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 1A10
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias 72 kDa gelatinase, CLG4, CLG4A72 kDa type IV collagenase, collagenase type IV-A, EC 3.4.24, EC 3.4.24.24, Gelatinase A, matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase), matrix metalloproteinase 2 (gelatinase A, 72kD gelatinase, 72kD type IVcollagenase), Matrix metalloproteinase-2, matrix metalloproteinase-II, MMP-2, MMP-II, MONA, neutrophil gelatinase, TBE-1matrix metalloproteinase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IVcollagenase)
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human MMP-2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, HIF Target Genes, Hypoxia, Metastasis, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4313
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.