Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-24/MT5-MMP Antibody (143908), HRP, Novus Biologicals™
SDP

Catalog No. NB174481 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174481 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174481 Supplier Novus Biologicals Supplier No. FAB924H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

MMP-24/MT5-MMP Monoclonal antibody specifically detects MMP-24/MT5-MMP in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen MMP-24/MT5-MMP
Applications Western Blot
Classification Monoclonal
Clone 143908
Conjugate HRP
Formulation PBS
Gene Alias EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 24 (membrane-inserted), matrix metalloproteinase 24 (membrane-inserted), matrix metalloproteinase-24, membrane-type 5 matrix metalloproteinase, Membrane-type matrix metalloproteinase 5, Membrane-type-5 matrix metalloproteinase, MMP-24, MT5MMP, MT5-MMPMMP25, MT-MMP 5, MTMMP5, MT-MMP5
Host Species Rat
Immunogen E. coli-derived recombinant mouse MMP-24/MT5-MMP, Arg125-Gln296, Accession # Q9R0S2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10893
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.