Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMP-24/MT5-MMP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | MMP-24/MT5-MMP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
MMP-24/MT5-MMP Polyclonal specifically detects MMP-24/MT5-MMP in Human samples. It is validated for Western Blot.Specifications
MMP-24/MT5-MMP | |
Polyclonal | |
Rabbit | |
Q9Y5R2 | |
10893 | |
Synthetic peptides corresponding to MMP24(matrix metallopeptidase 24 (membrane-inserted)) The peptide sequence was selected from the middle region of MMP24 (NP_006681). Peptide sequence GSCLPREGIDTALRWEPVGKTYFFKGERYWRYSEERRATDPGYPKPITVW. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 24 (membrane-inserted), matrix metalloproteinase 24 (membrane-inserted), matrix metalloproteinase-24, membrane-type 5 matrix metalloproteinase, Membrane-type matrix metalloproteinase 5, Membrane-type-5 matrix metalloproteinase, MMP-24, MT5MMP, MT5-MMPMMP25, MT-MMP 5, MTMMP5, MT-MMP5 | |
MMP24 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title