Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-24/MT5-MMP Antibody (RM0289-7J71), FITC, Novus Biologicals™
SDP

Catalog No. NBP211803F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211803F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211803F Supplier Novus Biologicals Supplier No. NBP211803F
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MMP-24/MT5-MMP Monoclonal specifically detects MMP-24/MT5-MMP in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen MMP-24/MT5-MMP
Applications Flow Cytometry
Classification Monoclonal
Clone RM0289-7J71
Conjugate FITC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias EC 3.4.24, EC 3.4.24.-, EC 3.4.24.80, matrix metallopeptidase 24 (membrane-inserted), matrix metalloproteinase 24 (membrane-inserted), matrix metalloproteinase-24, membrane-type 5 matrix metalloproteinase, Membrane-type matrix metalloproteinase 5, Membrane-type-5 matrix metalloproteinase, MMP-24, MT5MMP, MT5-MMPMMP25, MT-MMP 5, MTMMP5, MT-MMP5
Gene Symbols MMP24
Host Species Rat
Immunogen Mouse recombinant MMP-24/MT5-MMP (extracellular domain)
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10893
Test Specificity This antibody weakly cross reacts with mouse MMP3 and MMP-9.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.