Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-25/MT6-MMP Antibody (MM0029-2B5), HRP, Novus Biologicals™
SDP

Catalog No. NB11060995H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11060995H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11060995H Supplier Novus Biologicals Supplier No. NB11060995H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMP-25/MT6-MMP Monoclonal specifically detects MMP-25/MT6-MMP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MMP-25/MT6-MMP
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0029-2B5
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with No Preservative
Gene Alias leukolysin, matrix metallopeptidase 25, matrix metallopeptidase-like 1, matrix metalloproteinase 20, matrix metalloproteinase 25, matrix metalloproteinase-25, matrix metalloproteinase-like 1, membrane-type 6 matrix metalloproteinase, membrane-type matrix metalloproteinase 6, membrane-type-6 matrix metalloproteinase, MMP20, MMP20A, MMP-25, MMPL1, MT6MMP, MT6-MMP, MT-MMP 6, MTMMP6, MT-MMP6
Gene Symbols MMP25
Host Species Mouse
Immunogen Human recombinant MMP-25/MT6-MMP
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 64386
Test Specificity MMP25 (MM0029-2B5)
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.