Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-3 Antibody (50647), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB173999 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173999 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173999 Supplier Novus Biologicals Supplier No. FAB513W
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMP-3 Monoclonal antibody specifically detects MMP-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry

Specifications

Antigen MMP-3
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 50647
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human MMP-3, Tyr18-Cys477, Accession # P08254
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4314
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.