Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB259213 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259213 0.025 mL
NB10091878 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB259213 Supplier Novus Biologicals Supplier No. NB10091878SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 8 publications

MMP-3 Polyclonal specifically detects MMP-3 in Human, Mouse, Rat samples. It is validated for IF/IHC, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen MMP-3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500- 1:1000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:100
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. P08254, P28862
Gene Alias CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1
Gene Symbols MMP3
Host Species Rabbit
Immunogen A synthetic peptide made to an C-terminal portion of the human MMP3 protein (between residues 400-477) [UniProt P08254]
Molecular Weight of Antigen 53 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4314
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.