Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-3 Antibody (MMP3/1994R), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB391713 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB391713 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB391713 Supplier Novus Biologicals Supplier No. NBP308627JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MMP-3 Monoclonal antibody specifically detects MMP-3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen MMP-3
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone MMP3/1994R
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, stromelysin-1, transin-1
Host Species Rabbit
Immunogen Recombinant fragment (around aa 317-327) of human MMP-3 protein (Uniprot: P08254)
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 4314
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.