Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-7 Antibody (704202) [DyLight 755], Novus Biologicals™
SDP

Catalog No. NB375020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375020 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375020 Supplier Novus Biologicals Supplier No. FAB7170Z
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MMP-7 Monoclonal antibody specifically detects MMP-7 in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen MMP-7
Applications Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 704202
Conjugate DyLight 755
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.24, EC 3.4.24.23, matrilysin, matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MMP-7, Met1-Leu264, Accession # Q10738
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4316
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.