Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-7 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB337701 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337701 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337701 Supplier Novus Biologicals Supplier No. NBP199123MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MMP-7 Polyclonal antibody specifically detects MMP-7 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen MMP-7
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.24, EC 3.4.24.23, matrilysin, matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human MMP7 protein (between residues 250-300) [UniProt P09237]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4316
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.