Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-9 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB201922 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB201922 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB201922 Supplier Novus Biologicals Supplier No. NBP241233B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MMP-9 Polyclonal antibody specifically detects MMP-9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen MMP-9
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid peptide near the center of human MMP9. The immunogen is located within amino acids 260 - 310 of MMP9 . Amino Acid Squence: PSERLYTQDGNADGK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 4318
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.