Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP-9 Antibody (V6P3F11*A4), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP250515IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250515IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250515IR Supplier Novus Biologicals Supplier No. NBP250515IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMP-9 Monoclonal specifically detects MMP-9 in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen MMP-9
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone V6P3F11*A4
Conjugate DyLight 755
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP-9, type V collagenase
Gene Symbols MMP9
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptide KAQLIDRGYP (C-terminal sequence)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cellular Markers, Extracellular Matrix, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 4318
Target Species Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.