Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP20 Antibody, Novus Biologicals™
SDP

Catalog No. NBP16232220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP16232220 20 μL
NBP162322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP16232220 Supplier Novus Biologicals Supplier No. NBP16232220UL

Rabbit Polyclonal Antibody

MMP20 Polyclonal specifically detects MMP20 in Human samples. It is validated for Western Blot.

Specifications

Antigen MMP20
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. O60882
Gene Alias AI2A2, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.35, Enamel metalloproteinase, Enamelysin, matrix metallopeptidase 20, matrix metalloproteinase 20 (enamelysin), matrix metalloproteinase-20, MMP-20
Gene Symbols MMP20
Host Species Rabbit
Immunogen Synthetic peptides corresponding to MMP20(matrix metallopeptidase 20) The peptide sequence was selected from the middle region of MMP20. Peptide sequence AAVYLREPQKTLFFVGDEYYSYDERKRKMEKDYPKNTEEEFSGVNGQIDA.
Molecular Weight of Antigen 43 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9313
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.