Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMP23B Antibody, Novus Biologicals™
SDP

Catalog No. NBP217313 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP217313 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP217313 Supplier Novus Biologicals Supplier No. NBP217313
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MMP23B Polyclonal specifically detects MMP23B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MMP23B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. O75900
Gene Alias EC 3.4.24.-, Femalysin, matrix metallopeptidase 23B, matrix metalloproteinase 22, matrix metalloproteinase 23B, matrix metalloproteinase in the female reproductive tract, Matrix metalloproteinase-21, Matrix metalloproteinase-22, matrix metalloproteinase-23, MIFR, MIFR-1MMP22femalysin, MMP21, MMP-21, MMP-22, MMP-23, MMP23A
Gene Symbols MMP23B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MMP23B. The exact sequence is proprietary.
Molecular Weight of Antigen 44 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8510
Test Specificity Human. Species reactivity based on sequence identity: Bovine (96%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.