Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMR/CD206/Mannose Receptor Antibody (15-2), Novus Biologicals™
SDP

Catalog No. NB6001415 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6001415 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB6001415 Supplier Novus Biologicals Supplier No. NB6001415
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMR/CD206/Mannose Receptor Monoclonal specifically detects MMR/CD206/Mannose Receptor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen MMR/CD206/Mannose Receptor
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Functional Assay
Classification Monoclonal
Clone 15-2
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10-1:100, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:100, Functional
Gene Accession No. P22897
Gene Alias CD206, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen
Gene Symbols MRC1
Host Species Mouse
Immunogen Purified human mannose receptor from human placental tissue
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4360
Test Specificity Human. Not yet tested in other species.
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.