Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMR/CD206/Mannose Receptor Antibody (685641), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB313399 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313399 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313399 Supplier Novus Biologicals Supplier No. FAB25342PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MMR/CD206/Mannose Receptor Monoclonal specifically detects MMR/CD206/Mannose Receptor in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen MMR/CD206/Mannose Receptor
Applications Flow Cytometry
Classification Monoclonal
Clone 685641
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD206, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse MMR/CD206, Leu19-Lys1383 (Thr399Ala) and (Leu407Phe), Accession # P22897
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4360
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.