Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMR/CD206/Mannose Receptor Rabbit anti-Human, Clone: S06-5I1, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP314900100UL
This item is not returnable.
View return policy
Description
MMR/CD206/Mannose Receptor Monoclonal antibody specifically detects MMR/CD206/Mannose Receptor in Human samples. It is validated for Western BlotSpecifications
MMR/CD206/Mannose Receptor | |
Monoclonal | |
Unconjugated | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
S06-5I1 | |
Western Blot | |
CD206, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen | |
A synthetic peptide of human MMR/CD206/Mannose Receptor (Uniprot#: P22897) | |
100 μg | |
Immunology, Innate Immunity, Signal Transduction | |
4360 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction