Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MMR/CD206/Mannose Receptor Rabbit anti-Human, Clone: S06-5I1, Novus Biologicals™

Rabbit Monoclonal Antibody
$275.50 - $584.50
Specifications
Antigen | MMR/CD206/Mannose Receptor |
---|---|
Clone | S06-5I1 |
Dilution | Western Blot |
Applications | Western Blot |
Classification | Monoclonal |
Description
MMR/CD206/Mannose Receptor Monoclonal antibody specifically detects MMR/CD206/Mannose Receptor in Human samples. It is validated for Western BlotSpecifications
MMR/CD206/Mannose Receptor | |
Western Blot | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD206, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen | |
A synthetic peptide of human MMR/CD206/Mannose Receptor (Uniprot#: P22897) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S06-5I1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Immunology, Innate Immunity, Signal Transduction | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
4360 | |
IgG | |
Affinity purified | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title