Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MMS19 like protein Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP30379920UL

 View more versions of this product

Catalog No. NB095639


Only null left
Add to Cart

Description

Description

MMS19 like protein Polyclonal specifically detects MMS19 like protein in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

MMS19 like protein
Polyclonal
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
64210
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
FLJ34167, MET18, MET18 homolog, MMS19 nucleotide excision repair homolog (S. cerevisiae), S. cerevisiae)
Recombinant fusion protein containing a sequence corresponding to amino acids 761-1030 of human MMS19 like protein (NP_071757.4).
20 μL
Primary
Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.