Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30635150UL
Description
MN1 Polyclonal antibody specifically detects MN1 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
MN1 | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
dJ353E16.2, meningioma (disrupted in balanced translocation) 1, meningioma (translocation balanced), meningioma chromosome region, meningioma chromosome region 1, MGCR, MGCR1, MGCR1-PEN, probable tumor suppressor protein MN1 | |
Produced in rabbits immunized with E. coli-derived Human MN1 fragment. | |
50 μg | |
Cell Cycle and Replication | |
4330 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence | |
Unconjugated | |
PBS, pH7.0 | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction