Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MNAB Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$382.00 - $646.00

Specifications

Antigen MNAB
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB438661
SDP
View Documents
Novus Biologicals
NBP21321225UL
25ul
Each for $382.00
Only null left
Add to Cart
 
NBP213212
SDP
View Documents
Novus Biologicals
NBP213212
0.1 mL
Each for $646.00
Only null left
Add to Cart
 
Description

Description

MNAB Polyclonal specifically detects MNAB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

MNAB
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
FLJ20301, FLJ20713, membrane associated DNA binding protein, membrane-associated nucleic acid binding protein, Membrane-associated nucleic acid-binding protein, MGC52176, MNAB, ring finger and CCCH-type domains 2, RING finger and CCCH-type zinc finger domain-containing protein 2, ring finger and CCCH-type zinc finger domains 2, RNF164RING finger protein 164
RC3H2
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500
Polyclonal
Rabbit
Zinc Finger
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
54542
This antibody was developed against a recombinant protein corresponding to the amino acids: GAISRSSRTGYHTTDPVQATASQGSATKPISVSDYVPYVNAVDSRWSSYGNEATSSAHYVERDRFIVTDLSGHRKHSSTGDLLSLELQQAKSNSLLLQ
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.