Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MNDA Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501728848 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8848 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8848 Supplier Proteintech Group Inc Supplier No. 135761AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Until recently, immunological markers for myeloid cells have been lacking, especially those which identify different levels of cellular differentiation. This antibody stains early precursor and mature forms of human myeloid cells. This mAB reacts with ant

Until recently, immunological markers for myeloid cells have been lacking, especially those which identify different levels of cellular differentiation. This antibody stains early precursor and mature forms of human myeloid cells. This mAB reacts with antigenic determits present in normal myeloid cells and leukemias of similar derivation.

Specifications

Antigen MNDA
Applications Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MNDA
Gene Accession No. P41218
Gene Alias MNDA, PYHIN3
Gene Symbols MNDA
Host Species Rabbit
Immunogen MNDA Fusion Protein Ag4476
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4332
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.