Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MNDA Rabbit anti-Human, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$518.60

Specifications

Antigen MNDA
Concentration 0.22 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-172-8848
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
135761AP150UL
150 μL
Each of 1 for $518.60
Only null left
Add to Cart
 
Description

Description

Until recently, immunological markers for myeloid cells have been lacking, especially those which identify different levels of cellular differentiation. This antibody stains early precursor and mature forms of human myeloid cells. This mAB reacts with ant

Until recently, immunological markers for myeloid cells have been lacking, especially those which identify different levels of cellular differentiation. This antibody stains early precursor and mature forms of human myeloid cells. This mAB reacts with antigenic determits present in normal myeloid cells and leukemias of similar derivation.
Specifications

Specifications

MNDA
Western Blot
Unconjugated
Rabbit
Human
P41218
4332
MNDA Fusion Protein Ag4476
Primary
-20°C
MNDA
0.22 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
MNDA, PYHIN3
MNDA
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.