Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MNX1/HLXB9 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB325495 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325495 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325495 Supplier Novus Biologicals Supplier No. NBP299434JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MNX1/HLXB9 Polyclonal antibody specifically detects MNX1/HLXB9 in Mouse samples. It is validated for Western Blot

Specifications

Antigen MNX1/HLXB9
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias HB9, HLXB9motor neuron and pancreas homeobox protein 1, homeobox HB9, HOXHB9Homeobox protein HB9, motor neuron and pancreas homeobox 1, SCRA1homeo box HB9
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human MNX1/HLXB9
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Neuroscience, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 3110
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.