Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOB4A Antibody (OTI3G9), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N272744A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272744A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272744A405 Supplier Novus Biologicals Supplier No. NBP272744AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MOB4A Monoclonal specifically detects MOB4A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MOB4A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3G9
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias MATS2, MGC33910, Mob1 homolog 1A, MOB1, Mps One Binder kinase activator-like 1A (yeast), Mob1A, Mob1Bmps one binder kinase activator-like 1A, MOB4Amob1A, Protein Mob4A
Gene Symbols MOB1B
Host Species Mouse
Immunogen Full length recombinant protein of human MOB4A (NP_775739) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 92597.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.