Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOB4A/MOBKL1B Antibody (01), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB351582 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB351582 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB351582 Supplier Novus Biologicals Supplier No. NBP306512MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MOB4A/MOBKL1B Monoclonal antibody specifically detects MOB4A/MOBKL1B in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen MOB4A/MOBKL1B
Applications Immunocytochemistry
Classification Monoclonal
Clone 01
Conjugate mFluor Violet 610 SE
Dilution Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ATS2;MATS2;MOB kinase activator 1B;MOB1B;MOB4A;MOBKL1A
Host Species Mouse
Immunogen This antibody was obtained from a mouse immunized with purified, recombinant Human MOB4A/MOBKL1B (Uniprot#: Q7L9L4; Met1-Arg216).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92597
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.