Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOBKL2B Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB325816 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325816 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325816 Supplier Novus Biologicals Supplier No. NBP299376MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MOBKL2B Polyclonal antibody specifically detects MOBKL2B in Human samples. It is validated for Western Blot

Specifications

Antigen MOBKL2B
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Em:AL163192.1, FLJ13204, FLJ23916, Mob1 homolog 2b, MOB1, Mps One Binder kinase activator-like 2B (yeast), MOB3BMGC32960, monopolar spindle 1 binding, MOB1, domain containing, mps one binder kinase activator-like 2B, Protein Mob3B
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MOBKL2B.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79817
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.