Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOBKL2B Antibody (OTI6C6), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP272748A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272748A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272748A70 Supplier Novus Biologicals Supplier No. NBP272748AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MOBKL2B Monoclonal specifically detects MOBKL2B in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MOBKL2B
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI6C6
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Em:AL163192.1, FLJ13204, FLJ23916, Mob1 homolog 2b, MOB1, Mps One Binder kinase activator-like 2B (yeast), MOB3BMGC32960, monopolar spindle 1 binding, MOB1, domain containing, mps one binder kinase activator-like 2B, Protein Mob3B
Gene Symbols MOB3B
Host Species Mouse
Immunogen Full length recombinant protein of human MOBKL2B (NP_079037) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 79817.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.