Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOBKL3 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB063840 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB063840 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB063840 Supplier Novus Biologicals Supplier No. NBP299981AF647

Rabbit Polyclonal Antibody

MOBKL3 Polyclonal specifically detects MOBKL3 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen MOBKL3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias 2C4D, CGI-95, Class II mMOB1, MOB1, MOB1, Mps One Binder kinase activator-like 3 (yeast), MOB1DKFZP564M112, Mob3, MOB3MGC12264, MOB4 MOB family member 4, phocein, MOBKL3, phocein, PHOCN, PREI3, PREI3mps one binder kinase activator-like 3, Preimplantation protein 3Mob1 homolog 3,2C4DCGI-95
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant MOBKL3 (Q9Y3A3-1; Val 2-Ala 225).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25843
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.