Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOCS2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP181045 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP181045 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP181045 Supplier Novus Biologicals Supplier No. NBP181045
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MOCS2 Polyclonal specifically detects MOCS2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MOCS2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:1000-1:2500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. O96033
Gene Alias EC 2.-, MCBPE, MOCO1-A, MOCO1-B, MOCO1MOCS2B, MOCS2A, molybdenum cofactor biosynthesis protein E, molybdenum cofactor synthesis 2, Molybdenum cofactor synthesis protein 2 large subunit, Molybdenum cofactor synthesis protein 2 small subunit, Molybdenum cofactor synthesis protein 2A, Molybdenum cofactor synthesis protein 2B, molybdopterin synthase catalytic subunit, molybdopterin synthase sulfur carrier subunit, Molybdopterin-synthase large subunit, Molybdopterin-synthase small subunit, MPT synthase large subunit, MPTS, Sulfur carrier protein MOCS2A
Gene Symbols MOCS2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:ISSSCFSLETKLPLSPPLVEDSAFEPSRKDMDEVEEKSKDVINFTAEKLSVDEVSQLVISPLCGAISL
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4338
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.